Gene Transcripts

Gene Symbol
GPR87
Entrez Gene ID
53836
Gene Name
G protein-coupled receptor 87
Gene Aliases
FKSG78, GPR95, KPG_002
Chromosome Location
Chr. 3: 151294086 - 151316848 on Build GRCh38
UniGene ID
Hs.591292
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023915.3 NP_076404.3 3 676
GenBank mRNA AB031548.1 - 3 608
AF237763.1 - 3 564
AF345566.1 - 1 354
AF411114.1 - 2 315
AK027784.1 - 3 676
BC009540.1 - 1 122
BC112941.1 - 3 585 4500655
BC117565.1 - 3 585 4521950
BC132888.1 - 3 642 40146910
BC132890.1 - 3 642 40146912
EU883576.1 - 2 315

View Full Product Details