Gene Transcripts

Gene Symbol
Klra6
Entrez Gene ID
16637
Gene Name
killer cell lectin-like receptor, subfamily A, member 6
Gene Aliases
Ly49f, ly-49f
Chromosome Location
Chr. 6: 130013033 - 130026960 on Build GRCm38
UniGene ID
Mm.14112
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008464.2 NP_032490.1 6 709
GenBank mRNA U10092.1 - 6 709

View Full Product Details