Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008786.2 | NP_032812.2 | 5 | 838 | |
GenBank mRNA | AK006162.1 | - | 5 | 516 | |
AK075632.1 | - | 4 | 325 | ||
AK148789.1 | - | 5 | 863 | ||
BC040750.1 | - | 5 | 431 | ||
BC049613.1 | - | 5 | 542 | ||
BC058966.1 | - | 5 | 527 | 5717963 | |
D38023.1 | - | 5 | 502 | ||
M60320.1 | - | 5 | 502 |