Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008983.2 | NP_033009.1 | 9 | 1764 | |
GenBank mRNA | AK019870.1 | - | 9 | 1664 | |
AK052275.1 | - | 9 | 1718 | ||
AK083006.1 | - | 9 | 1728 | ||
BC138376.1 | - | 9 | 1730 | 8861396 | |
BC145261.1 | - | 9 | 1730 |