Gene Transcripts
- Gene Symbol
- Rbpjl
- Entrez Gene ID
- 19668
- Gene Name
- recombination signal binding protein for immunoglobulin kappa J region-like
- Gene Aliases
- RBP-L, Rbpsuhl
- Chromosome Location
- Chr. 2: 164403056 - 164415448 on Build GRCm38
- UniGene ID
- Mm.473794
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_009036.1 |
NP_033062.1
|
12 |
2184 |
|
GenBank mRNA |
Y10926.1 |
- |
12 |
2184 |
|
View Full Product Details