Gene Transcripts

Gene Symbol
Sva
Entrez Gene ID
20939
Gene Name
seminal vesicle antigen
Gene Aliases
Chromosome Location
Chr. 6: 42038394 - 42042851 on Build GRCm38
UniGene ID
Mm.4119
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009299.2 NP_033325.2 2, 3 264
GenBank mRNA BC104304.1 - 2, 3 262
BC104305.1 - 2, 3 264 40043713
CJ055470.1 - 2, 3 264
M94179.1 - 2, 3 250

View Full Product Details