Gene Transcripts
- Gene Symbol
- Srpk2
- Entrez Gene ID
- 20817
- Gene Name
- serine/arginine-rich protein specific kinase 2
- Gene Aliases
- AW226533, AW492537, AW547358, Wbp6, mSRPK2
- Chromosome Location
- Chr. 5: 23503356 - 23684641 on Build GRCm38
- UniGene ID
- Mm.288728
- Species
- Mus musculus
- Gene Symbol
- Asic2
- Entrez Gene ID
- 11418
- Gene Name
- acid-sensing (proton-gated) ion channel 2
- Gene Aliases
- ACIC2, Accn1, BNC1, BNaC1, BNaC1a, Mdeg
- Chromosome Location
- Chr. 11: 80880163 - 81968396 on Build GRCm38
- UniGene ID
- Mm.234998
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
EF410096.1 |
- |
|
272 |
|
View Full Product Details