Gene Transcripts

Gene Symbol
Dynlt1b
Entrez Gene ID
21648
Gene Name
dynein light chain Tctex-type 1B
Gene Aliases
AGS2, Dynlt1, Tctex-1, Tctex1
Chromosome Location
Chr. 17: 6430112 - 6436296 on Build GRCm38
UniGene ID
Mm.1948
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009342.2 NP_033368.1 5 583
GenBank mRNA AK004520.1 - 336
AK133788.1 - 5 513
AK153971.1 - 5 518
BC043018.1 - 5 583 5684255
BC089472.1 - 4 381 6703917
M25825.1 - 495

Gene Symbol
Dynlt1a
Entrez Gene ID
100310872
Gene Name
dynein light chain Tctex-type 1A
Gene Aliases
Chromosome Location
Chr. 17: 6306344 - 6317474 on Build GRCm38
UniGene ID
Mm.381893
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166629.2 NP_001160101.1 5 557
GenBank mRNA AK030569.1 - 5 519
BC087868.1 - 5 557 6740486

Gene Symbol
Dynlt1c
Entrez Gene ID
100040563
Gene Name
dynein light chain Tctex-type 1C
Gene Aliases
100040603, Dynlt1d
Chromosome Location
Chr. 17: 6601779 - 6607891 on Build GRCm38
UniGene ID
Mm.457381
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166630.1 NP_001160102.1 5 514
GenBank mRNA AK168478.1 - 5 515

Gene Symbol
Dynlt1f
Entrez Gene ID
100040531
Gene Name
dynein light chain Tctex-type 1F
Gene Aliases
100040631, Dynlt1e
Chromosome Location
Chr. 17: 6646606 - 6655939 on Build GRCm38
UniGene ID
Mm.457987
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166627.1 NP_001160099.1 5 622
GenBank mRNA AA033150.1 - 1 185
AK014409.1 - 3 1189

View Full Product Details