Gene Transcripts

Gene Symbol
Prap1
Entrez Gene ID
22264
Gene Name
proline-rich acidic protein 1
Gene Aliases
Upa
Chromosome Location
Chr. 7: 140093396 - 140097203 on Build GRCm38
UniGene ID
Mm.439707
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009475.2 NP_033501.2 3 167
GenBank mRNA AA060392.1 - 3 165
BC051092.1 - 3 155 6529097
U28486.1 - 3 166

View Full Product Details