Gene Transcripts

Gene Symbol
Parp2
Entrez Gene ID
11546
Gene Name
poly (ADP-ribose) polymerase family, member 2
Gene Aliases
ARTD2, Adprt2, Adprtl2, Aspartl2, C78626, PARP-2
Chromosome Location
Chr. 14: 50807898 - 50821301 on Build GRCm38
UniGene ID
Mm.281482
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009632.2 NP_033762.1 2 174
GenBank mRNA AF072521.1 - 2 149
AJ007780.1 - 2 187
AK076193.1 - 2 201
AK141876.1 - 2 196
BC052000.1 - 2 174
BC062150.1 - 2 174 6333875

View Full Product Details