Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009948.2 | NP_034078.2 | 2 | 170 | |
GenBank mRNA | AB010826.1 | - | 1 | 105 | |
AF017174.2 | - | 2 | 170 | ||
AK029141.1 | - | 2 | 148 | ||
AK146851.1 | - | 2 | 172 | ||
BC018270.1 | - | 2 | 137 | 4920625 | |
CJ092541.1 | - | 2 | 170 |