Gene Transcripts

Gene Symbol
Ccr2
Entrez Gene ID
12772
Gene Name
chemokine (C-C motif) receptor 2
Gene Aliases
Cc-ckr-2, Ccr2a, Ccr2b, Ckr2, Ckr2a, Ckr2b, Cmkbr2, mJe-r
Chromosome Location
Chr. 9: 124101918 - 124109140 on Build GRCm38
UniGene ID
Mm.6272
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009915.2 NP_034045.1 3 2288
GenBank mRNA AK039755.1 - 1 1138
AK138599.1 - 2 2601
AK162705.1 - 1 1294
U56819.1 - 3 2289

View Full Product Details