Gene Transcripts

Gene Symbol
Dok1
Entrez Gene ID
13448
Gene Name
docking protein 1
Gene Aliases
AW557123, p62DOK
Chromosome Location
Chr. 6: 83030932 - 83033471 on Build GRCm38
UniGene ID
Mm.156
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291799.1 NP_001278728.1 3, 4 501
NM_010070.4 NP_034200.4 3, 4 503
GenBank mRNA AA790529.1 - 1, 2 38
AK136382.1 - 3, 4 504
AK169871.1 - 3, 4 499
BC013066.1 - 3, 4 497 3498170
CJ053718.1 - 3, 4 504
U78818.2 - 3, 4 468

View Full Product Details