Gene Transcripts

Gene Symbol
Mmp3
Entrez Gene ID
17392
Gene Name
matrix metallopeptidase 3
Gene Aliases
SLN-1, SLN1, STR-1, Stmy1, Str1
Chromosome Location
Chr. 9: 7445822 - 7455975 on Build GRCm38
UniGene ID
Mm.4993
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010809.1 NP_034939.1 8, 9 1319
GenBank mRNA AK148467.1 - 8, 9 1297
BC006725.1 - 8, 9 1283 3962288
X63162.1 - 8, 9 1319
X66402.1 - 8, 9 1261

View Full Product Details