Gene Transcripts

Gene Symbol
TAAR8
Entrez Gene ID
83551
Gene Name
trace amine associated receptor 8
Gene Aliases
GPR102, TA5, TAR5, TRAR5, TaR-5, TaR-8
Chromosome Location
Chr. 6: 132552693 - 132553721 on Build GRCh38
UniGene ID
Hs.350571
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053278.1 NP_444508.1 1 1001
GenBank mRNA AY183468.1 - 1 1001
BC069166.1 - 1 1001 7217002

View Full Product Details