Gene Transcripts
- Gene Symbol
- Numbl
- Entrez Gene ID
- 18223
- Gene Name
- numb-like
- Gene Aliases
- nbl
- Chromosome Location
- Chr. 7: 27258438 - 27282150 on Build GRCm38
- UniGene ID
- Mm.458153
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
U96441.1 |
- |
6 |
498 |
|
View Full Product Details