Gene Transcripts

Gene Symbol
Pcbp2
Entrez Gene ID
18521
Gene Name
poly(rC) binding protein 2
Gene Aliases
AW412548, Hnrpx, alphaCP-2
Chromosome Location
Chr. 15: 102470222 - 102500059 on Build GRCm38
UniGene ID
Mm.236513
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001103165.1 NP_001096635.1 4 350
NM_001103166.1 NP_001096636.1 4 350
NM_001174073.1 NP_001167544.1 4 326
NM_011042.2 NP_035172.2 4 350
GenBank mRNA AK012297.1 - 4 198
AK161527.1 - 4 174
AK169268.1 - 4 444
BC020478.1 - 3 95
BQ900329.1 - 4 326
EU604548.1 - 3 105
L19661.1 - 4 237

View Full Product Details