Gene Transcripts

Gene Symbol
MS4A2
Entrez Gene ID
2206
Gene Name
membrane spanning 4-domains A2
Gene Aliases
APY, ATOPY, FCER1B, FCERI, IGEL, IGER, IGHER, MS4A1
Chromosome Location
Chr. 11: 60088261 - 60098467 on Build GRCh38
UniGene ID
Hs.386748
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000139.4 NP_000130.1 2 203
NM_001256916.1 NP_001243845.1 2 203
XM_005273846.4 XP_005273903.1 3 518
XM_011544850.2 XP_011543152.1 3 136
XM_017017362.1 XP_016872851.1 3 518
GenBank mRNA AK304487.1 - 2 203
AK310142.1 - 2 203
BC074800.2 - 2 132 30915525
BC074843.2 - 2 132 30915222
D10583.1 - 2 151
DA972757.1 - 2 203
JF411082.1 - 2 101

View Full Product Details