Gene Transcripts

Gene Symbol
Tacc3
Entrez Gene ID
21335
Gene Name
transforming, acidic coiled-coil containing protein 3
Gene Aliases
Aint, C86661, Eric1
Chromosome Location
Chr. 5: 33658147 - 33672203 on Build GRCm38
UniGene ID
Mm.379024
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040435.2 NP_001035525.1 2, 3 737
GenBank mRNA AF093542.1 - 2, 3 238
AF156934.1 - 2, 3 288
AF203445.1 - 2, 3 238
AF203446.1 - 2, 3 238
AF247674.1 - 2, 3 238
AK145556.1 - 2, 3 298
AK160291.1 - 2, 3 299
BC003252.1 - 2, 3 273 3500413
BC055071.1 - 2713
BC066184.1 - 2, 3 273 30457053
BC068314.1 - 2, 3 273 30461783

View Full Product Details