Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011655.5 | NP_035785.1 | 3 | 491 | |
GenBank mRNA | AK010960.1 | - | 3 | 423 | |
AK011263.1 | - | 3 | 419 | ||
AK011765.1 | - | 3 | 423 | ||
AK012350.1 | - | 3 | 423 | ||
AK012365.1 | - | 1 | 62 | ||
AK013283.1 | - | 1 | 52 | ||
AK017806.1 | - | 3 | 423 | ||
AK051164.1 | - | 3 | 423 | ||
AK051393.1 | - | 3 | 425 | ||
AK076196.1 | - | 3 | 418 | ||
AK083327.1 | - | 3 | 423 | ||
AK146567.1 | - | 3 | 423 | ||
AK148978.1 | - | 3 | 418 | ||
AK151215.1 | - | 3 | 423 | ||
AK151670.1 | - | 3 | 423 | ||
AK160225.1 | - | 3 | 423 | ||
AK161575.1 | - | 3 | 423 | ||
AK165249.1 | - | 3 | 421 | ||
AK167691.1 | - | 3 | 423 | ||
AK167779.1 | - | 3 | 423 | ||
AK167808.1 | - | 3 | 418 | ||
AK167926.1 | - | 3 | 423 | ||
AK168271.1 | - | 3 | 418 | ||
AK169123.1 | - | 3 | 418 | ||
AK169210.1 | - | 3 | 422 | ||
AK169295.1 | - | 3 | 423 | ||
AK169606.1 | - | 3 | 424 | ||
AK188622.1 | - | 2 | 145 | ||
AK190170.1 | - | 1 | 107 | ||
AK209234.1 | - | 2 | 145 | ||
BC003825.1 | - | 3 | 397 | 3601177 | |
X04663.1 | - | 3 | 321 |