Gene Transcripts

Gene Symbol
Hsf4
Entrez Gene ID
26386
Gene Name
heat shock transcription factor 4
Gene Aliases
HSF 4, HSTF 4, HSTF4, ldis1, mHSF4
Chromosome Location
Chr. 8: 105269748 - 105275845 on Build GRCm38
UniGene ID
Mm.89204
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256042.1 NP_001242971.1 4 507
NM_001256044.1 NP_001242973.1 3 398
NM_011939.3 NP_036069.1 4 507
GenBank mRNA AB029349.1 - 4 439
AB029350.1 - 4 434
AF160966.1 - 4 434
AK148056.1 - 3 370
AK189360.1 - 1 80
AK216507.1 - 1 80
BC138130.1 - 4 507 8861149
BC145153.1 - 4 507
BC145154.1 - 4 507

View Full Product Details