Gene Transcripts

Gene Symbol
Ing1
Entrez Gene ID
26356
Gene Name
inhibitor of growth family, member 1
Gene Aliases
2610028J21Rik, AA407184, AI875420, mING1h, p33Ing1, p37Ing1b
Chromosome Location
Chr. 8: 11555762 - 11563251 on Build GRCm38
UniGene ID
Mm.25709
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB221554.1 - 2 545
AF149820.1 - 2 539
AF177755.1 - 2 527
AF177756.1 - 2 512
AF177757.1 - 2 1385
AK011594.1 - 1 511
AK035490.1 - 2 542
AK076482.1 - 2 435
AK141466.1 - 2 435
AK145553.1 - 2 437
AK157424.1 - 2 534
BC016573.1 - 2 927
BC147770.1 - 2 637 9087493
BC147784.1 - 2 637 9087503
CK624100.1 - 2 423
DV655694.1 - 2 505

View Full Product Details