Gene Transcripts
- Gene Symbol
- Cul1
- Entrez Gene ID
- 26965
- Gene Name
- cullin 1
- Gene Aliases
- Chromosome Location
- Chr. 6: 47452273 - 47526139 on Build GRCm38
- UniGene ID
- Mm.87611
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012042.3 |
NP_036172.1
|
3, 4 |
644 |
|
GenBank mRNA |
AF083216.1 |
- |
2, 3 |
368 |
|
AF136441.1 |
- |
2, 3 |
499 |
|
AF176910.1 |
- |
3, 4 |
533 |
|
AK147004.1 |
- |
3, 4 |
626 |
|
AK164274.1 |
- |
3, 4 |
611 |
|
AK165978.1 |
- |
3, 4 |
584 |
|
AK208612.1 |
- |
1, 2 |
137 |
|
BC004836.1 |
- |
2, 3 |
453 |
|
BC029260.1 |
- |
3, 4 |
644 |
2812400 |
View Full Product Details