Gene Transcripts

Gene Symbol
Pla2g2f
Entrez Gene ID
26971
Gene Name
phospholipase A2, group IIF
Gene Aliases
Chromosome Location
Chr. 4: 138750533 - 138757598 on Build GRCm38
UniGene ID
Mm.331989
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012045.4 NP_036175.2 3 349
GenBank mRNA AF166099.2 - 3 348
AK029254.1 - 3 339
BC125567.1 - 3 250 40130082

View Full Product Details