Gene Transcripts

Gene Symbol
Ctsj
Entrez Gene ID
26898
Gene Name
cathepsin J
Gene Aliases
CATP, CatRLP, Ctsp
Chromosome Location
Chr. 13: 61000277 - 61005958 on Build GRCm38
UniGene ID
Mm.31948
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012007.1 NP_036137.1 5 531
GenBank mRNA AF136272.1 - 5 531
AF158182.1 - 4 471
AK005526.1 - 5 538
AK131661.1 - 5 538
BC103769.1 - 5 527 30787117

View Full Product Details