Gene Transcripts

Gene Symbol
Fcer2a
Entrez Gene ID
14128
Gene Name
Fc receptor, IgE, low affinity II, alpha polypeptide
Gene Aliases
CD23, Fce2, Fcer2, Ly-42
Chromosome Location
Chr. 8: 3680726 - 3694174 on Build GRCm38
UniGene ID
Mm.1233
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001253737.1 NP_001240666.1 4, 5 297
NM_001253739.1 NP_001240668.1 4, 5 308
NM_001253743.1 NP_001240672.1 5, 6 357
NM_001253747.1 NP_001240676.1 4, 5 318
NM_013517.4 NP_038545.1 5, 6 438
GenBank mRNA AK156654.1 - 5, 6 441
AK172591.1 - 5, 6 441
BC119164.1 - 4, 5 305 8733913
BC119190.1 - 4, 5 305 8733939
M34163.1 - 5, 6 440
M99371.1 - 5, 6 430
S67072.1 - 4, 5 289
S67073.1 - 3, 4 169
S67074.1 - 4, 5 208
X64223.1 - 4, 5 297
X64224.1 - 4, 5 308

View Full Product Details