Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_017372.3 | NP_059068.1 | 1 | 113 | |
GenBank mRNA | AK148516.1 | - | 1 | 114 | |
AK150998.1 | - | 1 | 116 | ||
AK153244.1 | - | 1 | 116 | ||
AK153475.1 | - | 1 | 114 | ||
AK159276.1 | - | 1 | 163 | ||
AK159640.1 | - | 1 | 110 | ||
BC002069.1 | - | 1 | 105 | 3593360 | |
BC054463.1 | - | 1 | 105 | 6514977 |