Gene Transcripts

Gene Symbol
Tmem37
Entrez Gene ID
170706
Gene Name
transmembrane protein 37
Gene Aliases
AI173373, AW540203, Pr, Pr1
Chromosome Location
Chr. 1: 120067377 - 120074069 on Build GRCm38
UniGene ID
Mm.24750
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019432.2 NP_062305.1 2 218
GenBank mRNA AJ272046.1 - 2 170
AK002424.1 - 2 237
AK050134.1 - 2 218
AK086093.1 - 2 223
BC024613.1 - 2 214 4215775

View Full Product Details