Gene Transcripts
- Gene Symbol
- Stx1b
- Entrez Gene ID
- 56216
- Gene Name
- syntaxin 1B
- Gene Aliases
- Stx1b2, Stx1bl, Syn1b
- Chromosome Location
- Chr. 7: 127803900 - 127824531 on Build GRCm38
- UniGene ID
- Mm.439759
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
D45207.1 |
- |
|
1331 |
|
View Full Product Details