Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019712.3 | NP_062686.1 | 2 | 202 | |
GenBank mRNA | AF140599.1 | - | 2 | 162 | |
AK003159.1 | - | 2 | 164 | ||
AK004114.1 | - | 2 | 166 | ||
AK005127.1 | - | 2 | 167 | ||
AK149259.1 | - | 1 | 1405 | ||
AK152012.1 | - | 2 | 159 | ||
AK159571.1 | - | 2 | 159 | ||
AK182752.1 | - | 2 | 96 | ||
AK183716.1 | - | 2 | 96 | ||
BC027396.1 | - | 2 | 151 | 4952242 | |
BC051473.1 | - | 2 | 157 | 1430400 | |
BC056992.1 | - | 2 | 145 | 6818827 | |
CJ082632.1 | - | 2 | 222 |