Gene Transcripts
- Gene Symbol
- Mtm1
- Entrez Gene ID
- 17772
- Gene Name
- X-linked myotubular myopathy gene 1
- Gene Aliases
- AF073996, Mtm, mKIAA4176
- Chromosome Location
- Chr. X: 71210767 - 71315413 on Build GRCm38
- UniGene ID
- Mm.274981
- Species
- Mus musculus
- Gene Symbol
- Mccc1
- Entrez Gene ID
- 72039
- Gene Name
- methylcrotonoyl-Coenzyme A carboxylase 1 (alpha)
- Gene Aliases
- 1810045E08Rik, 2310058B18Rik, MCCalpha, Mcca, R75106
- Chromosome Location
- Chr. 3: 35959296 - 36000678 on Build GRCm38
- UniGene ID
- Mm.249016
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK141040.1 |
- |
2 |
5460 |
|
View Full Product Details