Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001164733.1 | NP_001158205.1 | 8 | 1047 | |
NM_001164734.1 | NP_001158206.1 | 8 | 1057 | ||
NM_019939.2 | NP_064323.2 | 7 | 1015 | ||
GenBank mRNA | AF161181.1 | - | 7 | 879 | |
AF199009.1 | - | 7 | 1011 | ||
AF199010.1 | - | 8 | 1053 | ||
AK144434.1 | - | 8 | 1058 | ||
AK178596.1 | - | 2 | 134 | ||
BC042868.2 | - | 7 | 962 | ||
BC046421.1 | - | 7 | 962 | ||
BC069908.1 | - | 7 | 1004 | ||
BC106140.1 | - | 7 | 962 | ||
BC138666.1 | - | 7 | 873 | 8861687 | |
BC145363.1 | - | 8 | 915 | 9053921 | |
BC145364.1 | - | 8 | 905 | 9053922 |