Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001290315.1 | NP_001277244.1 | 2 | 141 | |
NM_020043.3 | NP_064427.2 | 2 | 141 | ||
GenBank mRNA | AB052620.1 | - | 2 | 141 | |
AB052621.1 | - | 2 | 141 | ||
AF176694.1 | - | 2 | 92 | ||
AK079073.1 | - | 2 | 106 | ||
AK082162.1 | - | 2 | 134 | ||
AK122535.1 | - | 70 | |||
AK143256.1 | - | 2 | 93 | ||
BB624536.1 | - | 2 | 132 | ||
BC126879.1 | - | 2 | 92 | ||
BC126880.1 | - | 2 | 92 |