Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_021502.2 | NP_067477.1 | 2, 3 | 238 | |
GenBank mRNA | AB030200.1 | - | 2, 3 | 231 | |
AK007526.1 | - | 2, 3 | 208 | ||
BC028502.1 | - | 2, 3 | 218 | 1366538 | |
BC114968.1 | - | 206 | |||
BY091059.1 | - | 2, 3 | 239 |