Gene Transcripts
- Gene Symbol
- Mknk1
- Entrez Gene ID
- 17346
- Gene Name
- MAP kinase-interacting serine/threonine kinase 1
- Gene Aliases
- 2410048M24Rik, Mnk1
- Chromosome Location
- Chr. 4: 115838984 - 115879256 on Build GRCm38
- UniGene ID
- Mm.209327
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001285487.1 |
NP_001272416.1
|
6, 7 |
570 |
|
NM_001285488.1 |
NP_001272417.1
|
5, 6 |
464 |
|
NM_021461.5 |
NP_067436.1
|
4, 5 |
420 |
|
GenBank mRNA |
AK010697.1 |
- |
4, 5 |
398 |
|
AK141298.1 |
- |
4, 5 |
398 |
|
AK155934.1 |
- |
6, 7 |
531 |
|
AK169982.1 |
- |
5, 6 |
430 |
|
BC021369.1 |
- |
4, 5 |
376 |
5030044 |
BQ551007.1 |
- |
2, 3 |
127 |
|
Y11091.1 |
- |
4, 5 |
385 |
|
View Full Product Details