Gene Transcripts

Gene Symbol
Ncstn
Entrez Gene ID
59287
Gene Name
nicastrin
Gene Aliases
9430068N19Rik, AA727311, Aph2, D1Dau13e, Kiaa0253, Nct, mKIAA0253
Chromosome Location
Chr. 1: 172066013 - 172082749 on Build GRCm38
UniGene ID
Mm.218203
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021607.3 NP_067620.3 5, 6 642
GenBank mRNA AF240469.1 - 5, 6 592
AK033314.1 - 5, 6 585
AK129102.1 - 5, 6 642
AK170173.1 - 5, 6 588
AK170244.1 - 5, 6 607
AK170369.1 - 5, 6 631
AK172579.1 - 5, 6 587
BC019998.1 - 5, 6 587 4458780

View Full Product Details