Gene Transcripts

Gene Symbol
Qk
Entrez Gene ID
19317
Gene Name
quaking
Gene Aliases
1110003F05Rik, 1500005P18, QkI, l(17)-1Wis, l17Wis1
Chromosome Location
Chr. 17: 10202856 - 10319361 on Build GRCm38
UniGene ID
Mm.130694
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021881.2 NP_068681.1 7 2367
GenBank mRNA AF091047.1 - 3197
AK142236.1 - 2 980
U44940.1 - 7 2386

View Full Product Details