Gene Transcripts
- Gene Symbol
- Arpc1b
- Entrez Gene ID
- 11867
- Gene Name
- actin related protein 2/3 complex, subunit 1B
- Gene Aliases
- 41kDa, AA408064, AA408534, AA571392, AF007010, AW208418, L72, SOP2Hs, p41-ARC
- Chromosome Location
- Chr. 5: 145114256 - 145128186 on Build GRCm38
- UniGene ID
- Mm.30010
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_023142.2 |
NP_075631.2
|
5, 6 |
613 |
|
GenBank mRNA |
AF162768.1 |
- |
4, 5 |
538 |
|
AK002274.1 |
- |
5, 6 |
614 |
|
AK005378.1 |
- |
5, 6 |
614 |
|
AK147731.1 |
- |
5, 6 |
581 |
|
AK148145.1 |
- |
5, 6 |
607 |
|
AK151911.1 |
- |
5, 6 |
614 |
|
AK153510.1 |
- |
5, 6 |
614 |
|
AK157098.1 |
- |
5, 6 |
604 |
|
AK159371.1 |
- |
5, 6 |
612 |
|
AK159477.1 |
- |
5, 6 |
612 |
|
AK167978.1 |
- |
5, 6 |
543 |
|
AK168774.1 |
- |
5, 6 |
611 |
|
AK170759.1 |
- |
5, 6 |
603 |
|
AK171364.1 |
- |
5, 6 |
575 |
|
AK172124.1 |
- |
5, 6 |
581 |
|
AK178373.1 |
- |
1, 2 |
47 |
|
AK181008.1 |
- |
|
155 |
|
AK215938.1 |
- |
1, 2 |
47 |
|
BC003441.1 |
- |
5, 6 |
545 |
2650641 |
BC010275.1 |
- |
5, 6 |
543 |
3589768 |
BC020404.1 |
- |
2, 3 |
141 |
|
BC092051.1 |
- |
5, 6 |
557 |
3668682 |
View Full Product Details