Gene Transcripts

Gene Symbol
Sult1e1
Entrez Gene ID
20860
Gene Name
sulfotransferase family 1E, member 1
Gene Aliases
EST, ST1E1, Ste
Chromosome Location
Chr. 5: 87575968 - 87591611 on Build GRCm38
UniGene ID
Mm.89655
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023135.2 NP_075624.2 3 352
GenBank mRNA AK015750.1 - 3 336
BC034891.1 - 3 293 1479883
S78182.1 - 3 335

View Full Product Details