Gene Transcripts

Gene Symbol
KCNQ5
Entrez Gene ID
56479
Gene Name
potassium voltage-gated channel subfamily Q member 5
Gene Aliases
Kv7.5
Chromosome Location
Chr. 6: 72621843 - 73198851 on Build GRCh38
UniGene ID
Hs.445324
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001160130.1 NP_001153602.1 11 1942
NM_001160132.1 NP_001153604.1 12 1999
NM_001160133.1 NP_001153605.1 13 2026
NM_001160134.1 NP_001153606.1 10 1639
NM_019842.3 NP_062816.2 12 1969
GenBank mRNA AF202977.1 - 12 1517
AF249278.1 - 12 1731
AF263835.1 - 12 1409
AK299550.1 - 10 1239
AK299741.1 - 3 181
AK316070.1 - 3 183
BC117359.1 - 11 1613 40125910
BC143554.1 - 11 1613 9052065
BC143555.1 - 10 1486

View Full Product Details