Gene Transcripts

Gene Symbol
KCTD12
Entrez Gene ID
115207
Gene Name
potassium channel tetramerization domain containing 12
Gene Aliases
C13orf2, PFET1, PFETIN
Chromosome Location
Chr. 13: 76880169 - 76886405 on Build GRCh38
UniGene ID
Hs.644125
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138444.3 NP_612453.1 1 3073
GenBank mRNA AF359381.1 - 1 3058
AK057635.1 - 1 722
AK128363.1 - 2 2362
BC013764.1 - 1 2904 4521096

View Full Product Details