Gene Transcripts
- Gene Symbol
- Fam13c
- Entrez Gene ID
- 71721
- Gene Name
- family with sequence similarity 13, member C
- Gene Aliases
- 1200015N20Rik, C030038O19Rik, mKIAA1796
- Chromosome Location
- Chr. 10: 70440668 - 70558732 on Build GRCm38
- UniGene ID
- Mm.477813
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001143776.1 |
NP_001137248.1
|
3, 4 |
433 |
|
NM_001143777.1 |
NP_001137249.1
|
4, 5 |
537 |
|
NM_024244.4 |
NP_077206.3
|
3, 4 |
433 |
|
GenBank mRNA |
AK004797.1 |
- |
3, 4 |
417 |
|
AK021133.1 |
- |
4, 5 |
376 |
|
AK032053.1 |
- |
4, 5 |
349 |
|
AK036299.1 |
- |
4, 5 |
500 |
|
AK039513.1 |
- |
4, 5 |
501 |
|
AK041147.1 |
- |
3, 4 |
434 |
|
AK044049.1 |
- |
4, 5 |
460 |
|
AK081939.1 |
- |
4, 5 |
538 |
|
AK082202.1 |
- |
3, 4 |
429 |
|
AK220421.1 |
- |
4, 5 |
373 |
|
BC129839.1 |
- |
3, 4 |
389 |
40060810 |
BC129840.1 |
- |
3, 4 |
389 |
40060811 |
View Full Product Details