Gene Transcripts

Gene Symbol
Stx1b
Entrez Gene ID
56216
Gene Name
syntaxin 1B
Gene Aliases
Stx1b2, Stx1bl, Syn1b
Chromosome Location
Chr. 7: 127803900 - 127824531 on Build GRCm38
UniGene ID
Mm.439759
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024414.2 NP_077725.1 3 394
GenBank mRNA BC132042.1 - 3 189
BC132068.1 - 3 189 40130345
D29743.1 - 3 221

View Full Product Details