Gene Transcripts

Gene Symbol
Derl3
Entrez Gene ID
70377
Gene Name
Der1-like domain family, member 3
Gene Aliases
1810006I20Rik, 1810063P04Rik, IZP6, derlin-3
Chromosome Location
Chr. 10: 75893080 - 75895941 on Build GRCm38
UniGene ID
Mm.275878
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024440.2 NP_077760.1 7 1136
GenBank mRNA AB047555.1 - 7 1136
AK007348.1 - 7 997
AK007948.1 - 7 1122
AK172429.1 - 7 1122
BC004729.1 - 7 1119
BC019815.1 - 7 1115
BC099615.1 - 7 1116

View Full Product Details