Gene Transcripts

Gene Symbol
Nat9
Entrez Gene ID
66176
Gene Name
N-acetyltransferase 9 (GCN5-related, putative)
Gene Aliases
1110028N05Rik
Chromosome Location
Chr. 11: 115182831 - 115187886 on Build GRCm38
UniGene ID
Mm.86823
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025400.3 NP_079676.1 3 365
GenBank mRNA AK003948.1 - 3 319
AK009262.1 - 2 95
AK020083.1 - 3 345
AK082469.1 - 3 305
AK148048.1 - 3 397
AK154593.1 - 3 325
AK186542.1 - 3 205
BC011315.1 - 3 365 4013700

View Full Product Details