Gene Transcripts

Gene Symbol
Ufc1
Entrez Gene ID
66155
Gene Name
ubiquitin-fold modifier conjugating enzyme 1
Gene Aliases
1110021H02Rik, AA062310, ESTM29
Chromosome Location
Chr. 1: 171288564 - 171294982 on Build GRCm38
UniGene ID
Mm.2815
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025388.2 NP_079664.1 2 243
GenBank mRNA AK003070.1 - 1 145
AK003897.1 - 2 244
AK011260.1 - 1 134
BC021936.1 - 2 204 5124904

View Full Product Details