Gene Transcripts

Gene Symbol
FUT6
Entrez Gene ID
2528
Gene Name
fucosyltransferase 6
Gene Aliases
FCT3A, FT1A, Fuc-TVI, FucT-VI
Chromosome Location
Chr. 19: 5830626 - 5839753 on Build GRCh38
UniGene ID
Hs.631846
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000150.2 NP_000141.1 3 2238
NM_001040701.1 NP_001035791.1 2 2110
XM_005259526.4 XP_005259583.1 4 2305
XM_011527868.2 XP_011526170.1 4 2377
XM_011527869.2 XP_011526171.1 4 2374
XM_011527870.2 XP_011526172.1 5 2485
XM_011527872.2 XP_011526174.1 5 2505
XM_011527873.2 XP_011526175.1 4 2440
XM_011527874.2 XP_011526176.1 4 2460
XM_011527875.2 XP_011526177.1 5 2531
XM_011527876.2 XP_011526178.1 4 2486
XM_011527878.2 XP_011526180.1 4 2377
XM_011527879.2 XP_011526181.1 3 2332
GenBank mRNA AF131211.1 - 4 1319
AK098073.1 - 3 1095
AK225795.1 - 2 1617
AK313130.1 - 2 1169
BC061700.1 - 2 2100 4606746
M98825.1 - 1 1058
U27333.1 - 3 2196
U27334.1 - 3 1519
U27335.1 - 2 2071
U27336.1 - 2 1394
U27337.1 - 3 1681

View Full Product Details