Gene Transcripts

Gene Symbol
Fank1
Entrez Gene ID
66930
Gene Name
fibronectin type 3 and ankyrin repeat domains 1
Gene Aliases
1700007B22Rik, AI850911
Chromosome Location
Chr. 7: 133776856 - 133881536 on Build GRCm38
UniGene ID
Mm.87448
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025850.2 NP_080126.1 3 295
GenBank mRNA AK005694.1 - 3 305
AK017071.1 - 3 319
AY819025.1 - 3 325
BC046278.1 - 3 295 6310911

View Full Product Details