Gene Transcripts
- Gene Symbol
- Tspan31
- Entrez Gene ID
- 67125
- Gene Name
- tetraspanin 31
- Gene Aliases
- 2700085A14Rik, AW742554, Sas
- Chromosome Location
- Chr. 10: 127067290 - 127070261 on Build GRCm38
- UniGene ID
- Mm.35650
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_025982.4 |
NP_080258.1
|
3, 4 |
404 |
|
GenBank mRNA |
AK012457.1 |
- |
3, 4 |
405 |
|
AK012567.1 |
- |
3, 4 |
407 |
|
AK012853.1 |
- |
3, 4 |
407 |
|
AK013523.1 |
- |
3, 4 |
417 |
|
AK014183.1 |
- |
3, 4 |
403 |
|
AK075962.1 |
- |
3, 4 |
405 |
|
AK178759.1 |
- |
2, 3 |
142 |
|
AK180941.1 |
- |
2, 3 |
142 |
|
AK183488.1 |
- |
2, 3 |
120 |
|
AK188045.1 |
- |
2, 3 |
142 |
|
AK208118.1 |
- |
2, 3 |
118 |
|
BC023306.1 |
- |
3, 4 |
381 |
4165774 |
View Full Product Details