Gene Transcripts

Gene Symbol
Prl3a1
Entrez Gene ID
67000
Gene Name
prolactin family 3, subfamily a, member 1
Gene Aliases
1600016E11Rik, PLP-H, PLP-I, Plpi, Prlpi
Chromosome Location
Chr. 13: 27259489 - 27276660 on Build GRCm38
UniGene ID
Mm.69279
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025896.2 NP_080172.1 5 1211
GenBank mRNA AK005474.1 - 5 1212
BC100334.1 - 5 1148 30791042

View Full Product Details